Titanium(IV) chloride, 99.9%
Titanium(IV) chloride, 99.9%
Titanium(IV) chloride, 99.9%
Titanium(IV) chloride, 99.9%
Thermo Scientific Chemicals
Titanium(IV) chloride, 99.9%
CAS: 7550-45-0 | Cl4Ti | 189.667 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA197231000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point136.5°C
Chemical Name or MaterialTitanium(IV) chloride
Melting Point-24.0°C
CAS7550-45-0
ColorYellow
View more
Lewis acid
RUO – Research Use Only