Germanium tetrachloride, 99.99%, (trace metal basis)
Germanium tetrachloride, 99.99%, (trace metal basis)
Germanium tetrachloride, 99.99%, (trace metal basis)
Germanium tetrachloride, 99.99%, (trace metal basis)
Thermo Scientific Chemicals
Germanium tetrachloride, 99.99%, (trace metal basis)
CAS: 10038-98-9 | Cl4GeH8 | 222.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA197151000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point82.0°C to 84.0°C
Chemical Name or MaterialGermanium tetrachloride
Melting Point-49.5°C
Name Note99.99%
CAS10038-98-9
View more
for ultra trace analysis
RUO – Research Use Only