Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh
Thermo Scientific Chemicals
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh
CAS: 12055-62-8 | Ho2O3 | 377.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA197140500
Price (MYR)
2,636.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,636.00
EA
Add to cart
Specifications
Boiling Point3900.0°C
Chemical Name or MaterialHolmium(III) oxide
Melting Point2330.0°C
CAS12055-62-8
ColorBeige to Orange-Yellow
View more
for ultra trace analysis
RUO – Research Use Only