Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Thermo Scientific Chemicals
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Plastic bottle
Catalog number FSA197060500
Price (MYR)
2,829.00
EA
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,829.00
EA
Add to cart
Specifications
Boiling Point1412.0°C
Chemical Name or MaterialMagnesium chloride
Melting Point708.0°C
Name Note<10 mesh, anhydrous
CAS7786-30-3
View more
For ultra trace analysis
RUO – Research Use Only