Cadmium chloride hemipentahydrate, 99+%
Cadmium chloride hemipentahydrate, 99+%
Cadmium chloride hemipentahydrate, 99+%
Cadmium chloride hemipentahydrate, 99+%
Thermo Scientific Chemicals
Cadmium chloride hemipentahydrate, 99+%
CAS: 7790-78-5 | 2·5 H2O | 228.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA197022500
Price (MYR)
795.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
795.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCadmium chloride hemipentahydrate
Melting Point34.0°C
Name Notep.a.
CAS7790-78-5