Vanadium(III) chloride, 97%
Vanadium(III) chloride, 97%
Vanadium(III) chloride, 97%
Vanadium(III) chloride, 97%
Thermo Scientific Chemicals
Vanadium(III) chloride, 97%
Vanadium(III) chloride, 97%, Cl3V, CAS Number-7718-98-1, Vanadium trichloride, 100g, 96% min. (Argentometry), Black to Blue, 3.0000g/mL, 231-744-6, 13,289, 157.3, GHS Signal Word: Danger | CAS: 7718-98-1 | Cl3V | 157.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA197001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialVanadium(III) chloride
Melting Point250.0°C
Name Note99%
CAS7718-98-1
ColorBlack to Blue