Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
CAS: 8017-16-1 | (HO3P)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number FSA196950100
Price (MYR)
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyphosphoric acid
CAS8017-16-1
Boiling Point550.0°C
Density2.0500g/mL
Recommended StorageMay darken over time