Potassium iodide, 99%, extra pure, briquettes
Potassium iodide, 99%, extra pure, briquettes
Potassium iodide, 99%, extra pure, briquettes
Potassium iodide, 99%, extra pure, briquettes
Thermo Scientific Chemicals
Potassium iodide, 99%, extra pure, briquettes
CAS: 7681-11-0 | IK | 166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA196731000
Price (MYR)
473.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
473.00
EA
Add to cart
Specifications
Boiling Point1330.0°C
Chemical Name or MaterialPotassium iodide
Melting Point680.0°C
CAS7681-11-0