Copper(II) carbonate basic, pure, 54-56% Cu
Copper(II) carbonate basic, pure, 54-56% Cu
Copper(II) carbonate basic, pure, 54-56% Cu
Copper(II) carbonate basic, pure, 54-56% Cu
Copper(II) carbonate basic, pure, 54-56% Cu
Thermo Scientific Chemicals
Copper(II) carbonate basic, pure, 54-56% Cu
CAS: 12069-69-1 | CCuO3·CuO2H2 | 221.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA196675000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper(II) carbonate basic
Melting Point200.0°C
CAS12069-69-1
ColorGreen
Health Hazard 1GHS Signal Word: Warning
View more
Suitable for battery materials development.
RUO – Research Use Only