Bromine, 99.6%, for analysis
Bromine, 99.6%, for analysis
Bromine, 99.6%, for analysis
Bromine, 99.6%, for analysis
Thermo Scientific Chemicals
Bromine, 99.6%, for analysis
CAS: 7726-95-6 | Br | 79.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number FSA196662500
Price (MYR)
784.00
BOT
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
784.00
BOT
Add to cart
Specifications
Boiling Point58.7°C
Chemical Name or MaterialBromine
Melting Point-7.2°C
Name Notep.a.
CAS7726-95-6
View more
C-X bond formation reagent for analysis
RUO – Research Use Only