Sodium sulfate, 99%, extra pure, anhydrous
Sodium sulfate, 99%, extra pure, anhydrous
Sodium sulfate, 99%, extra pure, anhydrous
Sodium sulfate, 99%, extra pure, anhydrous
Thermo Scientific Chemicals
Sodium sulfate, 99%, extra pure, anhydrous
CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number FSA196640250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Boiling Point>1700.0°C
Chemical Name or MaterialSodium sulfate
Melting Point884.0°C
CAS7757-82-6