Potassium borohydride, 98+%
Potassium borohydride, 98+%
Potassium borohydride, 98+%
Potassium borohydride, 98+%
Thermo Scientific Chemicals
Potassium borohydride, 98+%
CAS: 13762-51-1 | BH4K | 53.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA196555000
Price (MYR)
2,304.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,304.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium borohydride
Melting Point500.0°C
Name Note98%
CAS13762-51-1
ColorWhite
View more
Reduction reagent
RUO – Research Use Only