Silver standard solution, for AAS, 1 mg/ml Ag in 0.5N HNO3
Silver standard solution, for AAS, 1 mg/ml Ag in 0.5N HNO3
Silver standard solution, for AAS, 1 mg/ml Ag in 0.5N HNO3
Silver standard solution, for AAS, 1 mg/ml Ag in 0.5N HNO3
Thermo Scientific Chemicals
Silver standard solution, for AAS, 1 mg/ml Ag in 0.5N HNO3
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA196305000
Price (MYR)
683.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
683.00
BOT
Add to cart
Chemical Identifiers
CAS7761-88-8, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialSilver standard solution
Name Note1 mg/ml Ag in 0.5N HNO3
PackagingPlastic bottle
CAS7761-88-8
Health Hazard 1GHS Signal Word: Danger
View more
for atomic adsorption spectroscopy
RUO – Research Use Only