Silicon standard solution, for AAS, 1 mg/ml Si in 2% NaOH
CAS: 7440-21-3 | Si | 28.09 g/mol
Have Questions?
Catalog Number
Quantity
FSA196291000
100 mL
Catalog number FSA196291000
Price (MYR)
448.00
BOT
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
448.00
BOT
Add to cart
Chemical Identifiers
CAS7440-21-3
Specifications
Chemical Name or MaterialSilicon standard solution
Name Note1 mg/ml Si in 2% NaOH
PackagingPlastic bottle
CAS7440-21-3
Health Hazard 2GHS P Statement Causes severe skin burns and eye damage.