Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
Thermo Scientific Chemicals
Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA196285000
Price (MYR)
775.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
775.00
BOT
Add to cart
Chemical Identifiers
CAS7782-49-2, 7647-01-0, 7732-18-5
Specifications
Chemical Name or MaterialSelenium standard solution
Name Note1 mg/ml Se in 10% HCl
PackagingPlastic bottle
CAS7782-49-2
Health Hazard 1GHS Signal Word: Danger
View more
for atomic adsorption spectroscopy
RUO – Research Use Only