Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
Thermo Scientific Chemicals
Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA196075000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS10099-74-8, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialLead standard solution
Name Note1 mg/ml Pb in 2% HNO3
PackagingPlastic bottle
CAS10099-74-8
Health Hazard 1Danger
View more
For atomic adsorption spectroscopy
RUO – Research Use Only