Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3
Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3
Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3
Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3
Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3
CAS: 7440-70-2 | Ca | 40.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA195885000
Price (MYR)
359.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
359.00
BOT
Add to cart
Chemical Identifiers
CAS7440-70-2, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialCalcium standard solution
Name Note1 mg/ml Ca in 0.5N HNO3
PackagingPlastic bottle
CAS7440-70-2
Health Hazard 1GHS Signal Word: Danger
View more
for atomic adsorption spectroscopy
RUO – Research Use Only