Carbol Fuchsin, pure
Carbol Fuchsin, pure
Carbol Fuchsin, pure
Carbol Fuchsin, pure
Thermo Scientific Chemicals
Carbol Fuchsin, pure
CAS: 4197-24-4 | C21H22ClN3 | 351.878 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA195791000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCarbol Fuchsin
Name NoteA mixture of Basic Fuchsin and phenol.
CAS4197-24-4
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing genetic defects.