Aluminum chloride, 99%, extra pure, anhydrous, granules
Aluminum chloride, 99%, extra pure, anhydrous, granules
Aluminum chloride, 99%, extra pure, anhydrous, granules
Aluminum chloride, 99%, extra pure, anhydrous, granules
Thermo Scientific Chemicals
Aluminum chloride, 99%, extra pure, anhydrous, granules
CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA195785000
Price (MYR)
467.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
467.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAluminum chloride
Melting Point194.0°C
CAS7446-70-0
ColorYellow
Health Hazard 1GHS Signal Word: Danger