Evans Blue
Evans Blue
Evans Blue
Thermo Scientific Chemicals
Evans Blue
CAS: 314-13-6 | C34H24N6Na4O14S4 | 960.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA195550250
Price (MYR)
648.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
648.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEvans Blue
CAS314-13-6
Molecular FormulaC34H24N6Na4O14S4
MDL NumberMFCD00004021
SynonymC.I. 23860, Direct Blue 53