hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt
CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol
Have Questions?
Change view
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA195370050
Price (MYR)
3,452.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,452.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydrogen hexachloroplatinate(IV) hydrate
Melting Point60.0°C
Name Noteca. 40% Pt
CAS26023-84-7
Health Hazard 1GHS Signal Word: Danger