Platinum on activated carbon, 5% Pt
Platinum on activated carbon, 5% Pt
Platinum on activated carbon, 5% Pt
Platinum on activated carbon, 5% Pt
Thermo Scientific Chemicals
Platinum on activated carbon, 5% Pt
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA195230100
Price (MYR)
1,378.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,378.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPlatinum on activated carbon
ColorBlack
Name Note5% Pt
CAS7440-06-4
Quantity10 g