Palladium on activated carbon, 10% Pd, unreduced, dry
Palladium on activated carbon, 10% Pd, unreduced, dry
Palladium on activated carbon, 10% Pd, unreduced, dry
Palladium on activated carbon, 10% Pd, unreduced, dry
Thermo Scientific Chemicals
Palladium on activated carbon, 10% Pd, unreduced, dry
CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Packaging:
View all
Glass Bottle
Catalog number FSA195030500
Price (MYR)
13,927.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
13,927.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPalladium on Activated Carbon
ColorBlack
Name Note10% Pd
CAS7440-05-3
Health Hazard 1Warning