Diethylene glycol dibutyl ether, 99+%, pure
Diethylene glycol dibutyl ether, 99+%, pure
Diethylene glycol dibutyl ether, 99+%, pure
Diethylene glycol dibutyl ether, 99+%, pure
Thermo Scientific Chemicals
Diethylene glycol dibutyl ether, 99+%, pure
CAS: 112-73-2 | C12H26O3 | 218.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA194982500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylene glycol dibutyl ether
CAS112-73-2
Melting Point-60.0°C
Density0.8800g/mL
EINECS Number204-001-9