Sodium borodeuteride, 98 atom% D, for NMR
Sodium borodeuteride, 98 atom% D, for NMR
Sodium borodeuteride, 98 atom% D, for NMR
Sodium borodeuteride, 98 atom% D, for NMR
Thermo Scientific Chemicals
Sodium borodeuteride, 98 atom% D, for NMR
CAS: 15681-89-7 | BH4Na | 37.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass Bottle
Catalog number FSA194950050
Price (MYR)
2,762.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,762.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium borodeuteride
Deuterium (D)97.5% min.
PackagingGlass Bottle
Physical FormPowder and/or Chunks
Quantity5 g
View more
For NMR
RUO – Research Use Only