Ammonium metavanadate, 99.5%, for analysis
Ammonium metavanadate, 99.5%, for analysis
Ammonium metavanadate, 99.5%, for analysis
Ammonium metavanadate, 99.5%, for analysis
Thermo Scientific Chemicals
Ammonium metavanadate, 99.5%, for analysis
CAS: 7803-55-6 | H4NO3V | 116.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number FSA194912500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium metavanadate
Melting Point200.0°C
CAS7803-55-6
ColorGreen to White
Health Hazard 1GHS Signal Word: Danger