Dibutyltin dichloride, 96%
Dibutyltin dichloride, 96%
Dibutyltin dichloride, 96%
Dibutyltin dichloride, 96%
Thermo Scientific Chemicals
Dibutyltin dichloride, 96%
CAS: 683-18-1 | C8H18Cl2Sn | 303.842 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Glass bottle
Catalog number FSA194872500
Price (MYR)
953.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
953.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDibutyltin dichloride
CAS683-18-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Suspected of causing genetic defects. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Fatal if inhaled. Toxic if swallowed. May damage fertility. May damage the unborn child.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.