Hexamethyldisiloxane, 98+%
Hexamethyldisiloxane, 98+%
Hexamethyldisiloxane, 98+%
Hexamethyldisiloxane, 98+%
Thermo Scientific Chemicals
Hexamethyldisiloxane, 98+%
CAS: 107-46-0 | C6H18OSi2 | 162.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA194795000
Price (MYR)
710.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
710.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexamethyldisiloxane
CAS107-46-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
View more
Silylation reagent
RUO – Research Use Only