Titanium(IV) isopropoxide, 98+%
Titanium(IV) isopropoxide, 98+%
Titanium(IV) isopropoxide, 98+%
Titanium(IV) isopropoxide, 98+%
Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%
CAS: 546-68-9 | C12H28O4Ti | 284.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA194702500
Price (MYR)
409.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
409.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium(IV) isopropoxide
CAS546-68-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remov
View more
Lewis acid
RUO – Research Use Only