Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Thermo Scientific Chemicals
Ammonium sulfate, 99.9995%, (trace metal basis)
CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA194150500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium sulfate
Melting Point280.0°C
CAS7783-20-2
ColorYellow to Orange
Health Hazard 1GHS Signal Word: Warning
View more
For ultra trace analysis
RUO – Research Use Only