Strontium chloride hexahydrate, 99%, extra pure
Strontium chloride hexahydrate, 99%, extra pure
Strontium chloride hexahydrate, 99%, extra pure
Strontium chloride hexahydrate, 99%, extra pure
Thermo Scientific Chemicals
Strontium chloride hexahydrate, 99%, extra pure
CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA194122500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialStrontium chloride hexahydrate
Melting Point115.0°C
CAS10025-70-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger