Potassium iodide, 99.995%, (trace metal basis), extra pure
Potassium iodide, 99.995%, (trace metal basis), extra pure
Potassium iodide, 99.995%, (trace metal basis), extra pure
Potassium iodide, 99.995%, (trace metal basis), extra pure
Thermo Scientific Chemicals
Potassium iodide, 99.995%, (trace metal basis), extra pure
CAS: 7681-11-0 | IK | 166 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA19379050050 gGlass bottle
Catalog number FSA193790500
Price (MYR)
1,221.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,221.00
EA
Add to cart
Specifications
Boiling Point1330.0°C
Chemical Name or MaterialPotassium iodide
Melting Point680.0°C
CAS7681-11-0
ColorWhite
View more
For ultra trace analysis
RUO – Research Use Only