cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)
Thermo Scientific Chemicals
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)
CAS: 15663-27-1 | Cl2H6N2Pt | 300.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA193762500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialcis-Dichlorodiamineplatinum(II)
Melting Point277.0°C
Name Note99.99%
CAS15663-27-1
ColorWhite
View more
For ultra trace analysis
RUO – Research Use Only