Platinum, 99.999%, (trace metal basis), powder
Platinum, 99.999%, (trace metal basis), powder
Platinum, 99.999%, (trace metal basis), powder
Platinum, 99.999%, (trace metal basis), powder
Thermo Scientific Chemicals
Platinum, 99.999%, (trace metal basis), powder
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1937100101 gGlass bottle
Catalog number FSA193710010
Price (MYR)
3,024.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,024.00
EA
Add to cart
Specifications
Chemical Name or MaterialPlatinum
ColorGray
CAS7440-06-4
Melting Point1772.0°C
Boiling Point3827.0°C
View more
For ultra trace analysis
RUO – Research Use Only