Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA193705000
Price (MYR)
503.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
503.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium dihydrogen phosphate
Melting Point190.0°C
CAS7722-76-1
ColorWhite
Physical FormCrystalline Powder