Lead(II) nitrate, 99.999%, (trace metal basis)
Lead(II) nitrate, 99.999%, (trace metal basis)
Lead(II) nitrate, 99.999%, (trace metal basis)
Lead(II) nitrate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals
Lead(II) nitrate, 99.999%, (trace metal basis)
CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA19332010010 gGlass bottle
Catalog number FSA193320100
Price (MYR)
456.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
EA
Add to cart
Specifications
Chemical Name or MaterialLead(II) nitrate
Melting Point470.0°C
CAS10099-74-8
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
For ultra trace analysis
RUO – Research Use Only