Lead(II) chloride, 99%
Lead(II) chloride, 99%
Lead(II) chloride, 99%
Lead(II) chloride, 99%
Thermo Scientific Chemicals
Lead(II) chloride, 99%
CAS: 7758-95-4 | Cl2Pb | 278.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number FSA193312500
Price (MYR)
492.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
492.00
BOT
Add to cart
Specifications
Boiling Point950.0°C
Chemical Name or MaterialLead(II) chloride
Melting Point501.0°C
CAS7758-95-4