Indium(III) chloride, 99.995%, (trace metal basis)
Indium(III) chloride, 99.995%, (trace metal basis)
Indium(III) chloride, 99.995%, (trace metal basis)
Indium(III) chloride, 99.995%, (trace metal basis)
Thermo Scientific Chemicals
Indium(III) chloride, 99.995%, (trace metal basis)
CAS: 10025-82-8 | Cl3In | 221.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA193190500
Price (MYR)
2,318.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,318.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIndium(III) chloride
Melting Point586.0°C
CAS10025-82-8
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
For ultra trace analysis
RUO – Research Use Only