Indium, 99.999%, (trace metal basis), powder
Indium, 99.999%, (trace metal basis), powder
Indium, 99.999%, (trace metal basis), powder
Indium, 99.999%, (trace metal basis), powder
Thermo Scientific Chemicals
Indium, 99.999%, (trace metal basis), powder
CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA193180050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIndium
ColorGray
CAS7440-74-6
Melting Point156.0°C
Boiling Point2000.0°C
View more
for ultra trace analysis
RUO – Research Use Only