Indium(III) oxide, 99.9997%, (trace metal basis)
Indium(III) oxide, 99.9997%, (trace metal basis)
Indium(III) oxide, 99.9997%, (trace metal basis)
Indium(III) oxide, 99.9997%, (trace metal basis)
Thermo Scientific Chemicals
Indium(III) oxide, 99.9997%, (trace metal basis)
CAS: 1312-43-2 | In2O3 | 277.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA193170050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIndium(III) oxide
CAS1312-43-2
ColorYellow
Physical FormPowder
Quantity5 g
View more
for ultra trace analysis
RUO – Research Use Only