Germanium, 99.999%, (trace metal basis), powder
Germanium, 99.999%, (trace metal basis), powder
Germanium, 99.999%, (trace metal basis), powder
Germanium, 99.999%, (trace metal basis), powder
Thermo Scientific Chemicals
Germanium, 99.999%, (trace metal basis), powder
CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA193110500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point2830.0°C
Chemical Name or MaterialGermanium
Melting Point937.0°C
Name Note99.999%
CAS7440-56-4
View more
For ultra trace analysis
RUO – Research Use Only