Poly(ethylene glycol), average M.W. 1000
Poly(ethylene glycol), average M.W. 1000
Poly(ethylene glycol), average M.W. 1000
Poly(ethylene glycol), average M.W. 1000
Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 1000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
Plastic bottle
Catalog number FSA192250051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(ethylene glycol)
ColorWhite
Name NoteAverage M.W. 1000
CAS25322-68-3
Physical FormWaxy Solid