Poly(ethylene glycol), average M.W. 400
Poly(ethylene glycol), average M.W. 400
Poly(ethylene glycol), average M.W. 400
Poly(ethylene glycol), average M.W. 400
Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 400
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic drum
Catalog number FSA192230051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(ethylene glycol)
ColorColorless
CAS25322-68-3
Physical FormLiquid
Boiling Point>200.0°C