Poly(ethylene glycol), average M.W. 300
Poly(ethylene glycol), average M.W. 300
Poly(ethylene glycol), average M.W. 300
Poly(ethylene glycol), average M.W. 300
Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 300
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA192220025
Price (MYR)
834.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
834.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(ethylene glycol)
ColorClear liquid
CAS25322-68-3
Melting Point-15.0°C to +10.0°C
EINECS Number500-038-2