Poly(propylene glycol), average M.W. 425
Poly(propylene glycol), average M.W. 425
Poly(propylene glycol), average M.W. 425
Poly(propylene glycol), average M.W. 425
Thermo Scientific Chemicals
Poly(propylene glycol), average M.W. 425
CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
2.5 kg
Packaging:
Glass bottle
Catalog number FSA192155000
Price (MYR)
1,075.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,075.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(propylene glycol)
Name NoteAverage M.W. 425
CAS25322-69-4
Boiling Point200.0°C
Melting Point-25.0°C