Cesium bromide, 99.9%, pure
Cesium bromide, 99.9%, pure
Cesium bromide, 99.9%, pure
Cesium bromide, 99.9%, pure
Thermo Scientific Chemicals
Cesium bromide, 99.9%, pure
CAS: 7787-69-1 | BrCs | 212.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA192061000
Price (MYR)
2,261.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,261.00
BOT
Add to cart
Specifications
Boiling Point1300.0°C
Chemical Name or MaterialCesium bromide
Melting Point636.0°C
Name Note99.9%
CAS7787-69-1