Cesium carbonate, 99.5%, for analysis
Cesium carbonate, 99.5%, for analysis
Cesium carbonate, 99.5%, for analysis
Cesium carbonate, 99.5%, for analysis
Thermo Scientific Chemicals
Cesium carbonate, 99.5%, for analysis
CAS: 534-17-8 | CCs2O3 | 325.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA192045000
Price (MYR)
1,966.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,966.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCesium carbonate
Melting Point>400°C
Name Note99.5%
CAS534-17-8