Fluorescamine, pure
Fluorescamine, pure
Fluorescamine, pure
Thermo Scientific Chemicals
Fluorescamine, pure
CAS: 38183-12-9 | C17H10O4 | 278.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
25 mg
100 mg
Packaging:
Glass bottle
Catalog number FSA191675000
Price (MYR)
1,942.00
EA
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,942.00
EA
Add to cart
Specifications
Chemical Name or MaterialFluorescamine
Melting Point155.0°C to 157.0°C
CAS38183-12-9
Health Hazard 1Warning
Molecular FormulaC17H10O4