Fast Blue BB, 98%, pure
Fast Blue BB, 98%, pure
Fast Blue BB, 98%, pure
Fast Blue BB, 98%, pure
Thermo Scientific Chemicals
Fast Blue BB, 98%, pure
CAS: 120-00-3 | C17H20N2O3 | 300.358 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA191571000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFast blue BB
Melting Point98.0°C to 100.0°C
Name Note98%
CAS120-00-3
Health Hazard 2GHS P Statement
Fatal if swallowed.