2',7'-Dichlorofluorescein, pure
2',7'-Dichlorofluorescein, pure
2',7'-Dichlorofluorescein, pure
2',7'-Dichlorofluorescein, pure
Thermo Scientific Chemicals
2',7'-Dichlorofluorescein, pure
CAS: 76-54-0 | C20H10Cl2O5 | 401.195 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA191531000
Price (MYR)
5,060.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,060.00
EA
Add to cart
Specifications
Absorbance2000 (at lambda min.)
Chemical Name or Material2', 7'-Dichlorofluorescein
CAS76-54-0
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.